Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017919.2 | NP_060389.2 | 4 | 532 | |
XM_011518820.2 | XP_011517122.1 | 4 | 615 | ||
XM_011518821.2 | XP_011517123.1 | 4 | 863 | ||
XM_011518823.2 | XP_011517125.1 | 2 | 353 | ||
XM_017014875.1 | XP_016870364.1 | 5 | 709 | ||
GenBank mRNA | AK000658.1 | - | 4 | 481 | |
AK095262.1 | - | 4 | 1143 | ||
AK295949.1 | - | 4 | 511 | ||
AK311412.1 | - | 5 | 614 | ||
AL833620.1 | - | 4 | 1044 | ||
BC070269.1 | - | 4 | 475 | ||
BC101564.1 | - | 3 | 469 | 8069070 | |
BC101566.1 | - | 3 | 469 | 8069072 | |
BC111009.1 | - | 4 | 492 | 5768578 | |
BC143430.1 | - | 3 | 469 | ||
DC318495.1 | - | 4 | 533 |