Gene Transcripts

Gene Symbol
SLC35A5
Entrez Gene ID
55032
Gene Name
solute carrier family 35 member A5
Gene Aliases
Chromosome Location
Chr. 3: 112561709 - 112584439 on Build GRCh38
UniGene ID
Hs.237480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017945.3 NP_060415.1 6 791
XM_011512948.1 XP_011511250.1 6 641
GenBank mRNA AK000737.1 - 6 694
AK098839.1 - 6 704
AY358994.1 - 6 700
BC005207.1 - 6 753 3829330
BC010307.1 - 6 753 3450563
BC013046.1 - 6 695 3875085

View Full Product Details