Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042410.1 | NP_001035869.1 | 6 | 683 | |
NM_001282792.1 | NP_001269721.1 | 3 | 362 | ||
NM_018089.2 | NP_060559.2 | 6 | 756 | ||
XM_005246663.2 | XP_005246720.1 | 6 | 675 | ||
XM_011511392.2 | XP_011509694.1 | 6 | 756 | ||
XM_017004420.1 | XP_016859909.1 | 6 | 675 | ||
XM_017004421.1 | XP_016859910.1 | 6 | 756 | ||
GenBank mRNA | AF364318.1 | - | 6 | 610 | |
AK001277.1 | - | 6 | 693 | ||
AK023206.1 | - | 6 | 683 | ||
AK095304.1 | - | 6 | 620 | ||
AK098282.1 | - | 4 | 1737 | ||
AK225275.1 | - | 6 | 681 | ||
AK225309.1 | - | 6 | 657 | ||
AK303077.1 | - | 5 | 489 | ||
AK303537.1 | - | 3 | 281 | ||
BC000238.1 | - | 6 | 638 | 3351742 | |
BC008948.2 | - | 6 | 667 | 2959748 | |
BC028334.1 | - | 3 | 284 | ||
DQ412564.1 | - | 6 | 693 |