Gene Transcripts
- Gene Symbol
- CHRNA2
- Entrez Gene ID
- 1135
- Gene Name
- cholinergic receptor nicotinic alpha 2 subunit
- Gene Aliases
- Chromosome Location
- Chr. 8: 27459761 - 27479296 on Build GRCh38
- UniGene ID
- Hs.57718
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000742.3 |
NP_000733.2
|
4, 5 |
956 |
|
NM_001282455.1 |
NP_001269384.1
|
4, 5 |
911 |
|
XM_005273397.2 |
XP_005273454.1
|
4, 5 |
880 |
|
XM_006716282.1 |
XP_006716345.1
|
5, 6 |
867 |
|
XM_011544388.1 |
XP_011542690.1
|
5, 6 |
958 |
|
XM_011544389.1 |
XP_011542691.1
|
4, 5 |
956 |
|
XM_017013003.1 |
XP_016868492.1
|
4, 5 |
888 |
|
XM_017013004.1 |
XP_016868493.1
|
5, 6 |
822 |
|
GenBank mRNA |
AK296348.1 |
- |
4, 5 |
466 |
|
AK308465.1 |
- |
1 |
187 |
|
AK314787.1 |
- |
4, 5 |
874 |
|
BC153866.1 |
- |
3, 4 |
382 |
40121686 |
CD013901.1 |
- |
3, 4 |
522 |
|
DA096824.1 |
- |
3, 4 |
348 |
|
U62431.1 |
- |
4, 5 |
901 |
|
Y16281.1 |
- |
3, 4 |
347 |
|
View Full Product Details