Gene Transcripts

Gene Symbol
ASXL2
Entrez Gene ID
55252
Gene Name
additional sex combs like 2, transcriptional regulator
Gene Aliases
ASXH2
Chromosome Location
Chr. 2: 25733758 - 25878485 on Build GRCh38
UniGene ID
Hs.119815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018263.4 NP_060733.4 13 2811
XM_006712039.3 XP_006712102.1 9 4932
XM_006712040.2 XP_006712103.1 9 2218
XM_011532950.2 XP_011531252.1 12 2850
XM_011532951.2 XP_011531253.1 11 2538
XM_017004429.1 XP_016859918.1 13 2641
XM_017004430.1 XP_016859919.1 10 4998
GenBank mRNA AB051472.2 - 13 2811
AB084279.1 - 13 2674
BX647527.1 - 14 2884
BX648218.1 - 13 2836
CR933613.1 - 13 2855

View Full Product Details