Gene Transcripts

Gene Symbol
LOC105747689
Entrez Gene ID
105747689
Gene Name
uncharacterized LOC105747689
Gene Aliases
Chromosome Location
Chr. 2: 191690181 - 191710469 on Build GRCh38
UniGene ID
Hs.104972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK130181.1 - 332

Gene Symbol
SLC35C1
Entrez Gene ID
55343
Gene Name
solute carrier family 35 member C1
Gene Aliases
CDG2C, FUCT1
Chromosome Location
Chr. 11: 45804072 - 45813016 on Build GRCh38
UniGene ID
Hs.12211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145265.1 NP_001138737.1 2 610
NM_001145266.1 NP_001138738.1 2 513
NM_018389.4 NP_060859.4 1 884
XM_011520203.2 XP_011518505.1 1 884
GenBank mRNA AF323970.1 - 1 172
AF326199.1 - 1 781
AK002182.1 - 2 286
AK027394.1 - 1 861
AK027747.1 - 1 825
AK074929.1 - 1 885
AK091245.1 - 1 1568
AK225428.1 - 2 308
AK315473.1 - 1 586
AL547030.3 - 2 609
BC001427.2 - 1 189 3139599
DB287838.1 - 2 499

View Full Product Details