Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017518.7 | NP_059988.3 | 7, 8 | 775 | |
GenBank mRNA | AF267739.1 | - | 7, 8 | 706 | |
AK300650.1 | - | 7, 8 | 727 | ||
AL122036.1 | - | 3, 4 | 310 | ||
BC008141.1 | - | 7, 8 | 775 | 4154976 |