Gene Transcripts

Gene Symbol
MIOX
Entrez Gene ID
55586
Gene Name
myo-inositol oxygenase
Gene Aliases
ALDRL6
Chromosome Location
Chr. 22: 50483988 - 50490321 on Build GRCh38
UniGene ID
Hs.129227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017584.5 NP_060054.4 6, 7 626
XM_005261925.3 XP_005261982.1 5, 6 493
GenBank mRNA AF197129.1 - 5, 6 512
AY064416.1 - 6, 7 539
AY738258.1 - 5, 6 512
BC012025.1 - 6, 7 658 4454512
BC064395.1 - 6, 7 534
BC073848.1 - 6, 7 546 5186354
CR456478.1 - 6, 7 571

View Full Product Details