Gene Transcripts
- Gene Symbol
- FERMT1
- Entrez Gene ID
- 55612
- Gene Name
- fermitin family member 1
- Gene Aliases
- C20orf42, DTGCU2, KIND1, UNC112A, URP1
- Chromosome Location
- Chr. 20: 6074845 - 6123544 on Build GRCh38
- UniGene ID
- Hs.472054
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017671.4 |
NP_060141.3
|
13, 14 |
2507 |
|
GenBank mRNA |
AB105105.1 |
- |
13, 14 |
2507 |
|
AF443278.1 |
- |
13, 14 |
2076 |
|
AK000123.1 |
- |
5, 6 |
678 |
|
AK000747.1 |
- |
3, 4 |
373 |
|
AK225216.1 |
- |
5, 6 |
678 |
|
AK314394.1 |
- |
13, 14 |
1897 |
|
AY137240.1 |
- |
12, 13 |
1718 |
|
BC040545.1 |
- |
12, 13 |
1762 |
5277060 |
BX647465.1 |
- |
8, 9 |
914 |
|
View Full Product Details