Gene Transcripts
- Gene Symbol
- ZDHHC7
- Entrez Gene ID
- 55625
- Gene Name
- zinc finger DHHC-type containing 7
- Gene Aliases
- DHHC7, SERZ-B, SERZ1, ZNF370
- Chromosome Location
- Chr. 16: 84974460 - 85011732 on Build GRCh38
- UniGene ID
- Hs.461610
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK000286.1 |
- |
3 |
412 |
|
View Full Product Details