Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017864.3 | NP_060334.2 | 8 | 1037 | |
NR_073444.1 | 8 | 1037 | |||
XM_017013616.1 | XP_016869105.1 | 8 | 1037 | ||
XM_017013617.1 | XP_016869106.1 | 8 | 1037 | ||
GenBank mRNA | AB161944.1 | - | 8 | 1021 | |
AK091278.1 | - | 7 | 774 | ||
BC136754.1 | - | 8 | 951 | 9020744 | |
BC144396.1 | - | 8 | 951 | ||
BX538203.1 | - | 4 | 360 |