Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017902.2 | NP_060372.2 | 5 | 833 | |
XM_011539940.2 | XP_011538242.1 | 5 | 844 | ||
XM_011539941.1 | XP_011538243.1 | 5 | 711 | ||
GenBank mRNA | AF395830.1 | - | 5 | 768 | |
AK000622.1 | - | 5 | 778 | ||
AK295353.1 | - | 5 | 860 | ||
AL359615.1 | - | 4 | 496 | ||
BC007719.2 | - | 5 | 761 | 4138066 |