Gene Transcripts

Gene Symbol
H2AFJ
Entrez Gene ID
55766
Gene Name
H2A histone family member J
Gene Aliases
H2AJ
Chromosome Location
Chr. 12: 14774336 - 14778002 on Build GRCh38
UniGene ID
Hs.524280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177925.3 NP_808760.1 1 1346
NR_027716.2 2 1032
GenBank mRNA AK001765.1 - 2 792
AK303301.1 - 3 909
AL133626.1 - 2 786

View Full Product Details