Gene Transcripts

Gene Symbol
LMBRD1
Entrez Gene ID
55788
Gene Name
LMBR1 domain containing 1
Gene Aliases
C6orf209, LMBD1, MAHCF, NESI
Chromosome Location
Chr. 6: 69675749 - 69797157 on Build GRCh38
UniGene ID
Hs.271643
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018368.3 NP_060838.3 8, 9 1048
XM_011535941.1 XP_011534243.1 8, 9 898
GenBank mRNA AF113224.1 - 8, 9 768
AF208863.1 - 7, 8 896
AF211480.1 - 5, 6 386
AK002102.1 - 8, 9 797
AK225187.1 - 8, 9 797
AK225537.1 - 8, 9 782
AY136817.1 - 7, 8 553
BC010360.1 - 6, 7 614
BC047073.1 - 8, 9 887 5274221
BX647737.1 - 7, 8 640

View Full Product Details