Gene Transcripts

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
Chromosome Location
Chr. 14: 88385638 - 88470350 on Build GRCh38
UniGene ID
Hs.525518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040428.3 NP_001035518.1 5 852
NM_018418.4 NP_060888.2 6 948
XM_005267851.1 XP_005267908.1 6 919
XM_005267852.1 XP_005267909.1 5 848
XM_005267854.1 XP_005267911.1 7 981
XM_005267855.1 XP_005267912.1 6 914
XM_006720204.1 XP_006720267.1 6 919
XM_006720205.1 XP_006720268.1 6 919
XM_011536951.1 XP_011535253.1 6 1113
XM_011536952.1 XP_011535254.1 4 731
XM_011536953.1 XP_011535255.1 5 1052
XM_017021452.1 XP_016876941.1 6 1113
XM_017021453.1 XP_016876942.1 6 1079
XM_017021454.1 XP_016876943.1 7 981
XM_017021455.1 XP_016876944.1 6 914
XM_017021456.1 XP_016876945.1 7 927
XM_017021457.1 XP_016876946.1 5 1052
GenBank mRNA AF144487.3 - 6 928
AF144488.2 - 4 653
AK055864.1 - 6 870
AK225746.1 - 7 919
AK290631.1 - 5 823
AK293007.1 - 7 901
AL136604.1 - 5 832
BC008656.1 - 1 380
BC090875.1 - 6 1235 30383306
CR533563.1 - 5 677
CR936777.1 - 6 894
EU668353.1 - 6 924

View Full Product Details