Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136204.2 | NP_001129676.1 | 9 | 1303 | |
NM_018715.3 | NP_061185.1 | 10 | 1472 | ||
GenBank mRNA | AB040903.1 | - | 10 | 1403 | |
AJ421269.1 | - | 10 | 1489 | ||
AL359612.1 | - | 7 | 822 | ||
BC004933.1 | - | 6 | 743 | ||
BC042141.1 | - | 9 | 1268 | 5932549 | |
BC053908.1 | - | 9 | 1301 | 6502476 | |
BC142946.1 | - | 9 | 1059 |