Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173487.1 | NP_001166958.1 | 4 | 990 | |
NM_001173488.1 | NP_001166959.1 | 2 | 1376 | ||
NM_017544.3 | NP_060014.2 | 3 | 892 | ||
XM_011531365.2 | XP_011529667.1 | 4 | 1522 | ||
GenBank mRNA | AJ011812.2 | - | 2 | 1376 | |
AL539002.3 | - | 4 | 919 | ||
AY208891.1 | - | 3 | 892 | ||
BC040379.1 | - | 3 | 874 | 4777539 | |
BC047878.2 | - | 3 | 859 | 5228666 | |
BC068514.1 | - | 3 | 870 | 5299667 | |
EF165092.1 | - | 1 | 450 | ||
EF165094.1 | - | 1 | 95 | ||
EF216688.1 | - | 2 | 1376 | ||
EF216689.1 | - | 2 | 1376 |