Gene Transcripts
- Gene Symbol
- CFC1
- Entrez Gene ID
- 55997
- Gene Name
- cripto, FRL-1, cryptic family 1
- Gene Aliases
- CFC1B, CRYPTIC, DTGA2, HTX2
- Chromosome Location
- Chr. 2: 130592165 - 130599575 on Build GRCh38
- UniGene ID
- Hs.567542
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270420.1 |
NP_001257349.1
|
4, 5 |
648 |
|
NM_032545.3 |
NP_115934.1
|
5, 6 |
763 |
|
GenBank mRNA |
AF312769.1 |
- |
5, 6 |
521 |
|
AK290094.1 |
- |
5, 6 |
739 |
|
AK315326.1 |
- |
5, 6 |
763 |
|
BC069508.1 |
- |
5, 6 |
545 |
7262189 |
BC074825.2 |
- |
5, 6 |
524 |
30915323 |
BC074826.2 |
- |
5, 6 |
524 |
30915540 |
BC110080.1 |
- |
5, 6 |
636 |
40030641 |
BC146897.1 |
- |
5, 6 |
640 |
9056825 |
BG654700.1 |
- |
2, 3 |
219 |
|
CA941621.1 |
- |
4, 5 |
580 |
|
- Gene Symbol
- CFC1B
- Entrez Gene ID
- 653275
- Gene Name
- cripto, FRL-1, cryptic family 1B
- Gene Aliases
- Chromosome Location
- Chr. 2: 130521094 - 130527992 on Build GRCh38
- UniGene ID
- Hs.503733
- Species
- Homo sapiens
View Full Product Details