Gene Transcripts

Gene Symbol
SMG9
Entrez Gene ID
56006
Gene Name
SMG9, nonsense mediated mRNA decay factor
Gene Aliases
C19orf61, F17127_1, HBMS
Chromosome Location
Chr. 19: 43727992 - 43754990 on Build GRCh38
UniGene ID
Hs.466875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005259057.2 XP_005259114.1 3 521
XM_011527113.1 XP_011525415.1 3 276
XM_011527114.1 XP_011525416.1 3 214
XM_011527115.1 XP_011525417.1 3 282
XM_011527116.1 XP_011525418.1 3 408
GenBank mRNA AK022948.1 - 3 493
AK309830.1 - 3 398
AL136606.1 - 3 447
BC008869.2 - 3 433 4131601
DA356912.1 - 3 521

View Full Product Details