Gene Transcripts
- Gene Symbol
- KLHL4
- Entrez Gene ID
- 56062
- Gene Name
- kelch like family member 4
- Gene Aliases
- DKELCHL, KHL4
- Chromosome Location
- Chr. X: 87517541 - 87670050 on Build GRCh38
- UniGene ID
- Hs.49075
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019117.4 |
NP_061990.2
|
2, 3 |
773 |
|
NM_057162.2 |
NP_476503.1
|
2, 3 |
773 |
|
GenBank mRNA |
AB051474.1 |
- |
2, 3 |
719 |
|
AF284765.1 |
- |
2, 3 |
680 |
|
AF284766.1 |
- |
2, 3 |
680 |
|
AK022715.1 |
- |
2, 3 |
196 |
|
AK056457.1 |
- |
2, 3 |
741 |
|
AL832514.1 |
- |
2, 3 |
713 |
|
BC140839.1 |
- |
2, 3 |
642 |
9021707 |
BC140840.1 |
- |
2, 3 |
642 |
9021708 |
BC142652.1 |
- |
2, 3 |
619 |
40148044 |
BC146677.1 |
- |
2, 3 |
619 |
40148894 |
BC161675.1 |
- |
2, 3 |
719 |
8862604 |
View Full Product Details