Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019613.3 | NP_062559.2 | 3 | 388 | |
XM_017024862.1 | XP_016880351.1 | 3 | 303 | ||
XM_017024863.1 | XP_016880352.1 | 3 | 147 | ||
GenBank mRNA | AF091083.1 | - | 1 | 31 | |
AK297477.1 | - | 3 | 142 | ||
AM182326.1 | - | 3 | 193 | ||
AY691427.1 | - | 1 | 19 | ||
BC000974.2 | - | 3 | 171 | ||
BC007838.1 | - | 3 | 360 | ||
CN262716.1 | - | 3 | 388 | ||
CR456770.1 | - | 1 | 19 |