Gene Transcripts

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
Chromosome Location
Chr. 4: 103077618 - 103099867 on Build GRCh38
UniGene ID
Hs.124696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020139.3 NP_064524.3 4 280
XM_005263140.3 XP_005263197.1 4 275
XM_006714274.3 XP_006714337.1 4 275
XM_011532127.2 XP_011530429.1 4 317
XM_011532128.2 XP_011530430.1 4 317
XM_017008461.1 XP_016863950.1 4 314
XM_017008462.1 XP_016863951.1 4 314
GenBank mRNA AF164790.1 - 4 244
AK023323.1 - 4 235
AK290160.1 - 4 236
AK300626.1 - 4 125
AY358841.1 - 4 246
BC001953.1 - 4 273 2905279
BC037277.2 - 4 242 4838679
BC095414.1 - 4 248 5284436
BI544872.1 - 4 301
CR457309.1 - 3 175

View Full Product Details