Gene Transcripts

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
Chromosome Location
Chr. 4: 103077618 - 103099867 on Build GRCh38
UniGene ID
Hs.124696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020139.3 NP_064524.3 5 376
XM_005263140.3 XP_005263197.1 5 371
XM_006714274.3 XP_006714337.1 5 371
XM_011532127.2 XP_011530429.1 5 413
XM_011532128.2 XP_011530430.1 5 413
XM_017008461.1 XP_016863950.1 5 410
XM_017008462.1 XP_016863951.1 5 410
GenBank mRNA AF164790.1 - 5 340
AK023323.1 - 5 331
AK128463.1 - 6 1074
AK290160.1 - 5 332
AK291335.1 - 5 338
AK300626.1 - 5 221
AY358841.1 - 5 342
BC001953.1 - 5 369 2905279
BC037277.2 - 5 338 4838679
BC095414.1 - 5 344 5284436
BI544872.1 - 5 397
CR457309.1 - 4 271

View Full Product Details