Gene Transcripts
- Gene Symbol
- SPIRE1
- Entrez Gene ID
- 56907
- Gene Name
- spire type actin nucleation factor 1
- Gene Aliases
- Spir-1
- Chromosome Location
- Chr. 18: 12446512 - 12658102 on Build GRCh38
- UniGene ID
- Hs.515283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128626.1 |
NP_001122098.1
|
4, 5 |
774 |
|
NM_001128627.1 |
NP_001122099.1
|
4, 5 |
472 |
|
NM_020148.2 |
NP_064533.3
|
4, 5 |
774 |
|
XM_005258122.4 |
XP_005258179.1
|
4, 5 |
957 |
|
XM_011525701.1 |
XP_011524003.1
|
4, 5 |
963 |
|
XM_011525702.1 |
XP_011524004.1
|
4, 5 |
475 |
|
XM_011525703.2 |
XP_011524005.1
|
4, 5 |
957 |
|
GenBank mRNA |
AB032961.2 |
- |
4, 5 |
826 |
|
AJ277587.1 |
- |
2, 3 |
361 |
|
AK290180.1 |
- |
2, 3 |
332 |
|
AK301711.1 |
- |
3, 4 |
241 |
|
AL833817.1 |
- |
5, 6 |
452 |
|
BC115005.1 |
- |
4, 5 |
599 |
7782242 |
BC125206.1 |
- |
2, 3 |
288 |
40123468 |
BC125207.1 |
- |
2, 3 |
288 |
40123469 |
DB056921.1 |
- |
4, 5 |
472 |
|
View Full Product Details