Gene Transcripts
- Gene Symbol
- PARP6
- Entrez Gene ID
- 56965
- Gene Name
- poly(ADP-ribose) polymerase family member 6
- Gene Aliases
- ARTD17, PARP-6-B1, PARP-6-C, pART17
- Chromosome Location
- Chr. 15: 72241181 - 72272575 on Build GRCh38
- UniGene ID
- Hs.270244
- Species
- Homo sapiens
View Full Product Details