Gene Transcripts

Gene Symbol
CTNNBIP1
Entrez Gene ID
56998
Gene Name
catenin beta interacting protein 1
Gene Aliases
ICAT
Chromosome Location
Chr. 1: 9848276 - 9910322 on Build GRCh38
UniGene ID
Hs.463759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012329.1 NP_001012329.1 5 993
NM_020248.2 NP_064633.1 6 1027
XM_017001853.1 XP_016857342.1 6 1252
XM_017001854.1 XP_016857343.1 6 1126
XM_017001855.1 XP_016857344.1 5 969
GenBank mRNA AB021262.1 - 6 993
BC014300.1 - 5 981
BC146892.1 - 4 715 9056820
BC146896.1 - 4 715 9056824

View Full Product Details