Gene Transcripts

Gene Symbol
COQ9
Entrez Gene ID
57017
Gene Name
coenzyme Q9
Gene Aliases
C16orf49, COQ10D5
Chromosome Location
Chr. 16: 57447425 - 57461275 on Build GRCh38
UniGene ID
Hs.513632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020312.3 NP_064708.1 7 916
GenBank mRNA AF161444.1 - 7 850
AK290627.1 - 7 846
AL136884.1 - 2 350
BC001478.2 - 5 491
BC029341.1 - 7 850
BC054340.1 - 7 850 6514554
BC064946.1 - 7 851 6023461

View Full Product Details