Gene Transcripts

Gene Symbol
AGTRAP
Entrez Gene ID
57085
Gene Name
angiotensin II receptor associated protein
Gene Aliases
ATRAP
Chromosome Location
Chr. 1: 11736085 - 11750771 on Build GRCh38
UniGene ID
Hs.464438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040194.1 NP_001035284.1 5 911
NM_001040195.1 NP_001035285.1 4 876
NM_001040196.1 NP_001035286.1 6 1008
NM_001040197.1 NP_001035287.1 6 1029
NM_020350.4 NP_065083.3 5 932
XM_011541799.2 XP_011540101.2 6 925
XM_011541800.2 XP_011540102.1 6 946
XM_011541802.1 XP_011540104.1 7 1022
GenBank mRNA AF312374.1 - 5 863
AK290042.1 - 5 875
BC011946.1 - 5 817
BC017328.2 - 5 849 4546585
BQ672962.1 - 1 416

View Full Product Details