Gene Transcripts

Gene Symbol
LRTM1
Entrez Gene ID
57408
Gene Name
leucine rich repeats and transmembrane domains 1
Gene Aliases
HT017
Chromosome Location
Chr. 3: 54918333 - 54967088 on Build GRCh38
UniGene ID
Hs.591668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304389.1 NP_001291318.1 2 589
NM_020678.3 NP_065729.1 2 598
GenBank mRNA AF225421.1 - 2 569
BC040732.1 - 2 589 5223743
BC074741.2 - 2 459 30915199

View Full Product Details