Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242498.1 | NP_001229427.1 | 3 | 295 | |
NM_001242500.1 | NP_001229429.1 | 2 | 197 | ||
NM_020679.3 | NP_065730.2 | 3 | 274 | ||
XM_005257530.1 | XP_005257587.1 | 3 | 416 | ||
XM_011525051.2 | XP_011523353.1 | 3 | 511 | ||
XM_011525052.2 | XP_011523354.1 | 3 | 490 | ||
XM_011525054.2 | XP_011523356.1 | 3 | 511 | ||
XM_017024888.1 | XP_016880377.1 | 3 | 511 | ||
XM_017024889.1 | XP_016880378.1 | 3 | 295 | ||
XM_017024890.1 | XP_016880379.1 | 3 | 321 | ||
XM_017024891.1 | XP_016880380.1 | 3 | 233 | ||
XM_017024895.1 | XP_016880384.1 | 3 | 274 | ||
GenBank mRNA | AK300711.1 | - | 3 | 234 | |
BC033759.1 | - | 2 | 173 | 5215495 | |
BI752882.1 | - | 3 | 292 | ||
BU190758.1 | - | 2 | 196 | ||
BX404175.2 | - | 3 | 217 | ||
KU178731.1 | - | 2 | 137 | ||
KU178732.1 | - | 2 | 158 |