Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040456.1 | NP_001035546.1 | 2 | 286 | |
NM_001040457.1 | NP_001035547.1 | 3 | 408 | ||
XM_005250511.4 | XP_005250568.1 | 3 | 245 | ||
GenBank mRNA | AF226732.1 | - | 2 | 337 | |
AK026590.1 | - | 2 | 199 | ||
BC069017.1 | - | 2 | 237 | 3502576 | |
BG707286.1 | - | 2 | 297 | ||
DB050147.1 | - | 3 | 296 |