Gene Transcripts

Gene Symbol
TENM2
Entrez Gene ID
57451
Gene Name
teneurin transmembrane protein 2
Gene Aliases
ODZ2, TEN-M2, TNM2, ten-2
Chromosome Location
Chr. 5: 166979066 - 168264159 on Build GRCh38
UniGene ID
Hs.631957
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080428.2 NP_001073897.2 23 6230
NM_001122679.1 NP_001116151.1 27 6802
XM_005265950.2 XP_005266007.1 29 7449
XM_005265952.2 XP_005266009.1 25 6176
XM_006714897.2 XP_006714960.1 28 7428
XM_011534604.2 XP_011532906.1 28 7263
XM_017009660.1 XP_016865149.1 29 7476
XM_017009661.1 XP_016865150.1 28 7455
XM_017009662.1 XP_016865151.1 28 7266
XM_017009663.1 XP_016865152.1 26 6671
XM_017009664.1 XP_016865153.1 26 6584
XM_017009665.1 XP_016865154.1 26 6380
XM_017009666.1 XP_016865155.1 25 6359
XM_017009667.1 XP_016865156.1 26 6353
XM_017009668.1 XP_016865157.1 26 7890
XM_017009669.1 XP_016865158.1 26 7722
GenBank mRNA AB032953.2 - 18 4939
AK302302.1 - 1 367
AL137500.1 - 2 1055
BX648178.1 - 22 5824
CR936703.1 - 23 6163

View Full Product Details