Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160047.1 | NP_001153519.1 | 21 | 2810 | |
NM_020784.2 | NP_065835.2 | 21 | 2825 | ||
GenBank mRNA | AB037765.2 | - | 21 | 2737 | |
AK022563.1 | - | 7 | 1011 | ||
AK097748.1 | - | 22 | 2714 | ||
BC137081.1 | - | 21 | 2705 | 9021078 | |
BC137082.1 | - | 21 | 2705 | 9021079 | |
BC142650.1 | - | 21 | 2748 | 40148019 | |
BC144469.1 | - | 21 | 2690 | 9052996 | |
BC150205.1 | - | 21 | 2746 | 40148865 | |
BC152428.1 | - | 21 | 2737 | 8862542 |