Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001247987.1 | NP_001234916.1 | 4 | 637 | |
NM_020826.2 | NP_065877.1 | 2 | 419 | ||
GenBank mRNA | AB037848.1 | - | 2 | 405 | |
AK291009.1 | - | 2 | 340 | ||
AK308841.1 | - | 4 | 371 | ||
AL512743.1 | - | 2 | 342 | ||
BC093830.1 | - | 2 | 339 | 7939675 | |
BC093832.1 | - | 2 | 341 | 7939677 | |
BP251101.1 | - | 2 | 419 |