Gene Transcripts

Gene Symbol
LRCH2
Entrez Gene ID
57631
Gene Name
leucine rich repeats and calponin homology domain containing 2
Gene Aliases
dA204F4.4
Chromosome Location
Chr. X: 115110611 - 115234082 on Build GRCh38
UniGene ID
Hs.65366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243963.1 NP_001230892.1 7 1102
NM_020871.3 NP_065922.3 7 1102
XM_006724724.3 XP_006724787.2 7 1105
XM_017029696.1 XP_016885185.1 7 1111
XM_017029697.1 XP_016885186.1 7 1112
GenBank mRNA AB040928.1 - 3 283
BC125224.1 - 7 1095 40124466
BX647193.1 - 7 1102

View Full Product Details