Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032656.3 | NP_116045.2 | 7 | 1107 | |
XM_005253590.3 | XP_005253647.1 | 7 | 1111 | ||
XM_011538598.2 | XP_011536900.1 | 7 | 1111 | ||
XM_011538600.2 | XP_011536902.1 | 7 | 1111 | ||
GenBank mRNA | AB040950.1 | - | 4 | 479 | |
BC002575.2 | - | 4 | 490 | ||
BC037964.1 | - | 7 | 1089 | 5736582 |