Gene Transcripts

Gene Symbol
HMHB1
Entrez Gene ID
57824
Gene Name
histocompatibility minor HB-1
Gene Aliases
HB-1, HB-1Y, HLA-HB1
Chromosome Location
Chr. 5: 143812161 - 143820719 on Build GRCh38
UniGene ID
Hs.158320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021182.1 NP_067005.1 2 333
GenBank mRNA AF103884.1 - 2 333

View Full Product Details