Gene Transcripts

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
Chromosome Location
Chr. 20: 62952647 - 62968597 on Build GRCh38
UniGene ID
Hs.512686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302643.1 NP_001289572.1 6 863
NM_022082.3 NP_071365.3 5 801
XM_011528978.2 XP_011527280.1 5 713
GenBank mRNA AK074107.1 - 5 737
AK095473.1 - 5 652
AK125597.1 - 6 806
AK298503.1 - 5 707
BC025312.1 - 6 803 4563089
BC027447.1 - 1 22
BC038593.1 - 5 643

View Full Product Details