Gene Transcripts

Gene Symbol
DMRTB1
Entrez Gene ID
63948
Gene Name
DMRT like family B with proline rich C-terminal 1
Gene Aliases
Chromosome Location
Chr. 1: 53459399 - 53467488 on Build GRCh38
UniGene ID
Hs.131654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033067.2 NP_149056.1 1 179
GenBank mRNA AJ291671.1 - 1 106
AK057273.1 - 1 179

View Full Product Details