Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr. 11: 26189123 - 26663289 on Build GRCh38
UniGene ID
Hs.577269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001313726.1 NP_001300655.1 28 3065
NM_001313727.1 NP_001300656.1 24 2623
NM_031418.3 NP_113606.2 27 2973
XM_011520282.2 XP_011518584.1 14 1520
XM_017018118.1 XP_016873607.1 25 2601
XM_017018119.1 XP_016873608.1 22 2213
GenBank mRNA AJ300461.1 - 27 3680
AK295658.1 - 11 1008
AK295816.1 - 24 2535
AK302659.1 - 27 3012
AK304781.1 - 28 2906
AK316018.1 - 24 2535

View Full Product Details