Gene Transcripts

Gene Symbol
SPATA1
Entrez Gene ID
100505741
Gene Name
spermatogenesis associated 1
Gene Aliases
SP-2, SPAP1
Chromosome Location
Chr. 1: 84506301 - 84556495 on Build GRCh38
UniGene ID
Hs.584956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310156.1 NP_001297085.1 11 1293
XM_011542514.2 XP_011540816.1 10 1167
GenBank mRNA AF306347.1 - 11 1261
BG184610.1 - 2 196

View Full Product Details