Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000098.2 | NP_000089.1 | 3 | 764 | |
XM_005270484.1 | XP_005270541.1 | 3 | 432 | ||
GenBank mRNA | AK312687.1 | - | 3 | 368 | |
BC002445.2 | - | 3 | 373 | 3346025 | |
BC005172.2 | - | 3 | 373 | 3605012 | |
M58581.1 | - | 3 | 350 | ||
U09648.1 | - | 3 | 765 |