Gene Transcripts

Gene Symbol
EBF2
Entrez Gene ID
64641
Gene Name
early B-cell factor 2
Gene Aliases
COE2, EBF-2, O/E-3, OE-3
Chromosome Location
Chr. 8: 25841730 - 26045124 on Build GRCh38
UniGene ID
Hs.491292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022659.3 NP_073150.2 7 884
GenBank mRNA AK304372.1 - 6 532
AL701879.1 - 7 533

View Full Product Details