Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006849.2 | NP_006840.2 | 9, 10 | 1475 | |
GenBank mRNA | AK075485.1 | - | 10, 11 | 1392 | |
AK124931.1 | - | 8, 9 | 1494 | ||
AK295383.1 | - | 9, 10 | 1367 | ||
BC000537.2 | - | 9, 10 | 1409 | ||
BC075029.2 | - | 9, 10 | 1407 | ||
U19948.1 | - | 9, 10 | 1408 | ||
Z84717.1 | - | 5, 6 | 1703 |