Gene Transcripts

Gene Symbol
MARCH7
Entrez Gene ID
64844
Gene Name
membrane associated ring-CH-type finger 7
Gene Aliases
AXO, AXOT, MARCH-VII, RNF177
Chromosome Location
Chr. 2: 159712457 - 159768583 on Build GRCh38
UniGene ID
Hs.529272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282805.1 NP_001269734.1 10, 11 2291
NM_001282807.1 NP_001269736.1 7, 8 2038
NM_022826.3 NP_073737.1 8, 9 2163
XM_005246773.2 XP_005246830.1 8, 9 2186
XM_017004719.1 XP_016860208.1 7, 8 2021
XM_017004720.1 XP_016860209.1 7, 8 1983
GenBank mRNA AK022973.1 - 8, 9 2121
AK222880.1 - 8, 9 2113
AK292836.1 - 10, 11 2259
AK302347.1 - 7, 8 1987
AK316382.1 - 7, 8 1995
AL833651.1 - 4, 5 1187
BC003404.1 - 4, 5 562
BC065014.1 - 8, 9 2087 6169288

View Full Product Details