Gene Transcripts
- Gene Symbol
- INTS3
- Entrez Gene ID
- 65123
- Gene Name
- integrator complex subunit 3
- Gene Aliases
- C1orf193, C1orf60, INT3, SOSS-A, SOSSA
- Chromosome Location
- Chr. 1: 153728050 - 153774808 on Build GRCh38
- UniGene ID
- Hs.438723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001324475.1 |
NP_001311404.1
|
10, 11 |
1192 |
|
NM_023015.4 |
NP_075391.3
|
9, 10 |
1533 |
|
XM_005245459.2 |
XP_005245516.1
|
9, 10 |
1550 |
|
XM_005245461.2 |
XP_005245518.1
|
2, 3 |
133 |
|
XM_006711490.3 |
XP_006711553.1
|
10, 11 |
1003 |
|
XM_011509906.2 |
XP_011508208.1
|
9, 10 |
915 |
|
GenBank mRNA |
AK074979.1 |
- |
2, 3 |
231 |
|
AK290025.1 |
- |
10, 11 |
1186 |
|
AK304874.1 |
- |
3, 4 |
622 |
|
AL080172.1 |
- |
3, 4 |
342 |
|
AL832133.1 |
- |
8, 9 |
2568 |
|
BC105092.1 |
- |
9, 10 |
996 |
8144095 |
BC105094.1 |
- |
9, 10 |
996 |
8144097 |
BC116458.1 |
- |
9, 10 |
1528 |
5752962 |
BX640786.1 |
- |
9, 10 |
1519 |
|
BX640950.1 |
- |
9, 10 |
1534 |
|
CR627233.1 |
- |
7, 8 |
3066 |
|
CR749212.1 |
- |
7, 8 |
3060 |
|
CR749376.1 |
- |
8, 9 |
3185 |
|
View Full Product Details