Gene Transcripts

Gene Symbol
WNK3
Entrez Gene ID
65267
Gene Name
WNK lysine deficient protein kinase 3
Gene Aliases
PRKWNK3
Chromosome Location
Chr. X: 54192823 - 54358695 on Build GRCh38
UniGene ID
Hs.92423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002838.3 NP_001002838.1 4 1317
NM_020922.4 NP_065973.2 4 1317
XM_006724593.3 XP_006724656.1 4 1255
XM_011530802.2 XP_011529104.1 4 1255
XM_017029741.1 XP_016885230.1 4 1256
XM_017029742.1 XP_016885231.1 4 1333
XM_017029743.1 XP_016885232.1 4 1248
XM_017029744.1 XP_016885233.1 4 1150
XM_017029745.1 XP_016885234.1 4 1255
GenBank mRNA AJ409088.2 - 3 878
AY082340.1 - 3 878
AY352048.1 - 4 1317

View Full Product Details