Gene Transcripts

Gene Symbol
GGCT
Entrez Gene ID
79017
Gene Name
gamma-glutamylcyclotransferase
Gene Aliases
C7orf24, CRF21, GCTG, GGC
Chromosome Location
Chr. 7: 30496621 - 30504841 on Build GRCh38
UniGene ID
Hs.530024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024051.3 NP_076956.1 3 495
NR_037669.1 2 349
GenBank mRNA AK021779.1 - 2 974
AK315608.1 - 3 486
BC000625.2 - 3 490 3346822
BC013937.1 - 3 456 3997629
BC019243.1 - 3 487 3828006
BG527593.1 - 2 338

View Full Product Details