Gene Transcripts

Gene Symbol
C7orf25
Entrez Gene ID
79020
Gene Name
chromosome 7 open reading frame 25
Gene Aliases
Chromosome Location
Chr. 7: 42909273 - 42913161 on Build GRCh38
UniGene ID
Hs.698120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099858.1 NP_001093328.1 2 386
NM_024054.2 NP_076959.2 2 381
XM_005249844.3 XP_005249901.1 2 656
XM_017012596.1 XP_016868085.1 2 616
XM_017012597.1 XP_016868086.1 2 1148
GenBank mRNA AK024820.1 - 2 354
AK225194.1 - 2 360
AK298865.1 - 2 386
BC000769.2 - 2 345 2964418
BC001845.2 - 2 345 2964418
DC303669.1 - 2 366
DC366585.1 - 2 386

View Full Product Details