Gene Transcripts

Gene Symbol
NUP37
Entrez Gene ID
79023
Gene Name
nucleoporin 37
Gene Aliases
p37
Chromosome Location
Chr. 12: 102074189 - 102120139 on Build GRCh38
UniGene ID
Hs.444276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024057.3 NP_076962.2 7 860
GenBank mRNA AF514994.1 - 6 714
AK026271.1 - 6 695
BC000861.2 - 6 778 3459639

View Full Product Details